Key Documents
Z663581
Brady® thermal labeling system (TLS) labels
label for 50 mL conical tube, bottle, petri dish or deep well plate, W × L 38.10 mm × 19.05 mm, white Nylon
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
material
white Nylon
packaging
roll of 250 labels
manufacturer/tradename
Brady PTL-30-499
availability
available only in EU
W × L
38.10 mm × 19.05 mm
Legal Information
Brady is a registered trademark of Brady Worldwide, Inc.