28367-U
domnick hunter® LC-MS Nitrogen Generator
Model 12, 230 volt, output flow, 12 L/min
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41112500
NACRES:
SB.54
Recommended Products
feature
output flow 12 L/min
manufacturer/tradename
Domnick Hunter G4-510E
Looking for similar products? Visit Product Comparison Guide
General description
Nitrox LC-MS Nitrogen Generators are specially designed to meet the gas flow, purity, and pressure requirements of LC-MS. By providing a continuous supply of clean, dry, 99.5% pure nitrogen, they eliminate the need for expensive, hazardous, cumbersome, labor-intensive gas cylinders. Both models are compatible with atmospheric pressure chemical ionization (APCI) and electrospray ionization (ESI) interfaces.
These generators employ pressure swing adsorption technology to produce a continuous supply of high-purity nitrogen. An oil-free air compressor is integral to Model 12; use Model 18 with your current supply of oil-free compressed air. Either model delivers filtered compressed air (8.5 bar (125 psig)) to a bed of carbon molecular sieve, which selectively removes oxygen and other contaminants. Two adsorbent beds alternate between purification and regeneration modes, ensuring a continuous supply of nitrogen at specified purity levels.
CE approved.
Maintenance filter kits and warranty infomation available upon request. Contact Supelco Technical Service at [email protected] or phone 800-359-3041.
These generators employ pressure swing adsorption technology to produce a continuous supply of high-purity nitrogen. An oil-free air compressor is integral to Model 12; use Model 18 with your current supply of oil-free compressed air. Either model delivers filtered compressed air (8.5 bar (125 psig)) to a bed of carbon molecular sieve, which selectively removes oxygen and other contaminants. Two adsorbent beds alternate between purification and regeneration modes, ensuring a continuous supply of nitrogen at specified purity levels.
CE approved.
Maintenance filter kits and warranty infomation available upon request. Contact Supelco Technical Service at [email protected] or phone 800-359-3041.
Legal Information
domnick hunter is a registered trademark of Parker Hannifin
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service