Skip to Content
Merck
All Photos(1)

Key Documents

442869

Supelco

Tributyltin chloride

analytical standard

Synonym(s):

TBTC, Tributylchlorotin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SnCl
CAS Number:
Molecular Weight:
325.51
Beilstein:
3535715
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor pressure

<0.01 mmHg ( 20 °C)

CofA

current certificate can be downloaded

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.492 (lit.)

bp

171-173 °C/25 mmHg (lit.)

density

1.2 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

CCCC[Sn](Cl)(CCCC)CCCC

InChI

1S/3C4H9.ClH.Sn/c3*1-3-4-2;;/h3*1,3-4H2,2H3;1H;/q;;;;+1/p-1

InChI key

GCTFWCDSFPMHHS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tributyltin chloride is a salt derivative of tributyltin which is an organotin compound.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tributyltin chloride can be used as a precursor for the preparation of tributyltin (TBT) which may be used as a reference standard for the analysis of TBT in:
  • Seawater samples by gas chromatography-triple quadrupole mass spectrometry (GC-MS/MS) with electron ionization (EI) and multiple reaction monitoring (MRM) modes of operation.
  • Sediment samples by static headspace (SHS) GC combined with MS operating in selected ion monitoring (SIM) mode.
  • Environmental water sample matrices by stir bar sorptive extraction (SBSE) with sodium tetrahydridoborate (NaBH4)-based in-situ derivatization combined with large-volume injection (LVI) GC-MS operating under the SIM mode of detection.
  • Natural waters by LVI using programmed temperature vaporization (PTV) injector-GC combined with inductively coupled plasma (ICP) MS.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Development of a large volume injection method using a programmed temperature vaporization injector-gas chromatography hyphenated to ICP-MS for the simultaneous determination of mercury, tin and lead species at ultra-trace levels in natural waters.
Teran-Baamonde J, et al.
Journal of Chromatography A, 1547(3), 77-85 (2018)
Towards tributyltin quantification in natural water at the Environmental Quality Standard level required by the Water Framework Directive.
Alasonati E, et al.
Talanta, 160, 499-511 (2016)
Microbes a Tool for the Remediation of Organotin Pollution Determined by Static Headspace Gas Chromatography-Mass Spectrometry.
Finnegan C, et al.
Molecules (Basel), 23(3), 627-627 (2018)
Determination of tributyltin in environmental water matrices using stir bar sorptive extraction with in-situ derivatisation and large volume injection-gas chromatography-mass spectrometry.
Neng NR, et al.
Talanta, 126(3), 8-11 (2014)
Luca Amendola et al.
Analytica chimica acta, 975, 70-77 (2017-05-30)
Tributyltin (TBT) and pentachlorophenol (PCP) are listed as priority substances that warrant monitoring in the European Water Framework Directive (WFD 2000/60/CE), since TBT and PCP were identified as environmental endocrine disruptors and exhibit considerable toxicity toward both aquatic organisms and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service