Skip to Content
Merck
All Photos(1)

Key Documents

48614

Supelco

1,1,1-Trichloroethane solution

certified reference material, 200 μg/mL in methanol

Synonym(s):

Methyl chloroform

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

InChI

1S/C2H3Cl3/c1-2(3,4)5/h1H3

InChI key

UOCLXMDMGBRAIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,1,1-Trichloroethane is a dense and non-aqueous phase liquid, which can find wide applications as a solvent and stabilizer for vapor degreasing in industries.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 4-Dioxane drinking water occurrence data from the third unregulated contaminant monitoring rule
Adamson.TD, et al.
The Science of the Total Environment, 596, 236-245 (2017)
Combination of zero-valent iron and anaerobic microorganisms immobilized in luffa sponge for degrading 1, 1, 1-trichloroethane and the relevant microbial community analysis
Wang W and Wu Y
Applied Microbiology and Biotechnology, 101, 783-796 (2017)
K T Bogen et al.
Regulatory toxicology and pharmacology : RTP, 10(1), 26-50 (1989-08-01)
A methodology for using physiologically based pharmacokinetic (PBPK) models to derive predicted safe concentrations of noncarcinogens in drinking water for humans based on experimentally determined no observed adverse effect levels (NOAELs) in animals is presented and applied to the case
Mathieu Valcke et al.
Inhalation toxicology, 23(14), 863-877 (2011-11-17)
The objective of this study was to assess the impact of the exposure duration and intensity on the human kinetic adjustment factor (HKAF). A physiologically based pharmacokinetic model was used to compute target dose metrics (i.e. maximum blood concentration (C(max))
Ariel Grostern et al.
Environmental microbiology, 12(4), 1053-1060 (2010-01-22)
Chloroform (CF), or trichloromethane, is an ubiquitous environmental pollutant because of its widespread industrial use, historically poor disposal and recalcitrance to biodegradation. Chloroform is a potent inhibitor of metabolism and no known organism uses it as a growth substrate. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service