Skip to Content
Merck
All Photos(1)

Key Documents

57655-U

Ferrules for PEEK HPLC Fitting

10-32 ferrule

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

natural PEEK ferrule

packaging

pkg of 10 ea

fitting

10-32 ferrule

Looking for similar products? Visit Product Comparison Guide

Other Notes

R584790 --> material no.s 57655-U (10pk) and 58479 (5pk).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service