Skip to Content
Merck
α-Methyl-DL-tyrosine 98%

α-Methyl-DL-tyrosine

α-Methyl-DL-tyrosine 98%
Synonym(s):
DL-2-Methyl-3-(4-hydroxyphenyl)alanine, AMPT
Linear Formula:
4-(HO)C6H4CH2C(CH3)(NH2)CO2H
CAS Number:
Molecular Weight:
195.22
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Application
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing