- Zinc sensors with lower binding affinities for cellular imaging.
Zinc sensors with lower binding affinities for cellular imaging.
Dalton transactions (Cambridge, England : 2003) (2013-02-22)
Jin Hoon Kim, In Hong Hwang, Seung Pyo Jang, Juhye Kang, Sumi Kim, Insup Noh, Youngmee Kim, Cheal Kim, Roger G Harrison
PMID23426272
ABSTRACT
Zinc sensors based on 2,3-dipicolylamine (DPA) and quinoline have been synthesized. They fluoresced in the presence of Zn(2+) and remained fluorescent when other metal ions were present. Fluorescence enhancement of the sensors was not seen for most other metal ions. In vitro studies with fibroblasts showed fluorescence when sensor and Zn(2+) were present. As seen by single crystal X-ray analysis, four nitrogens from the sensor bind to Zn(2+). These new sensors have lower binding constants than the pentadentate sensors based on 2,2-DPA.