Skip to Content
Merck
All Photos(1)

Key Documents

AB10532

Sigma-Aldrich

Anti-GABA(A) receptor subunit gamma-2 Antibody

serum, from rabbit

Synonym(s):

gamma-aminobutyric acid (GABA) A receptor, gamma 2, gamma-aminobutyric acid A receptor, gamma 2, GABA(A) receptor subunit gamma-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
rat, human
technique(s):
western blot: suitable
citations:
1

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GABRG2(2566)

General description

GABA(A) receptor subunit gamma-2 is a protein belonging to a subclass of GABA receptors known as GABA (A). GABA (A) receptors are pentameric chloride channels composed of subunits belonging to different homology classes, i.e. α, β, γ, δ, θ, ε, π, and act as receptors for GABA, a crucial inhibitory neurotransmitter localized within the brain. GABA (A) gamma 2 subunit consists of 5 cysteine residues and carries the binding site for benzodiazepine. The subunit functions to promote receptor maintenance and clustering at the synapse, interacts with GABA receptor associated proteins such as GODZ and GABAWRAP, affects GABA (A) receptor conductance and kinetics, and is a contributing component to receptors participating in both phasic and tonic inhibition. Several mutations in the GABA(A) receptor subunit gamma-2 expression have been linked to epileptic syndromes that result in fibrile seizures.

Specificity

This antibody recognizes GABA(A) receptor subunit gamma-2.

Immunogen

GST-tagged recombinant protein corresponding to rat GABA(A) receptor subunit gamma-2.

Application

Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
This Anti-GABA(A) receptor subunit gamma-2 Antibody is validated for use in WB for the detection of GABA(A) receptor subunit gamma-2.

Quality

Evaluated by Western Blot in PC12 cell lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected GABA(A) receptor subunit gamma-2 on 10 µg of PC12 cell lysate.

Target description

~60 kDa observed

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
PC12 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gisele C Vaz et al.
Journal of neuroscience methods, 273, 55-63 (2016-10-25)
Liposomes are concentric lipid vesicles that allow a sustained release of entrapped substances. GABA (γ-aminobutyric acid) is the most prevalent inhibitory neurotransmitter in the central nervous system. Using GABA-containing liposomes (GL) prepared by the freeze-thawing method, we determined the effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service