Skip to Content
Merck
All Photos(1)

Key Documents

48905

Millipore

Giolitti Cantoni Broth

suitable for microbiology

Synonym(s):

Staphylococcus Enrichment Broth acc. to Giolitti and Cantoni

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

shelf life

limited shelf life, expiry date on the label

Quality Level

composition

casein peptone, 10 g/L
glycine, 1.2 g/L
lithium chloride, 5 g/L
D-mannitol, 20 g/L
meat extract, 5 g/L
sodium chloride, 5 g/L
sodium pyruvate, 3 g/L
yeast extract, 5 g/L

final pH

6.9±0.2 (25 °C)

application(s)

microbiology

suitability

selective and differential for Staphylococcus spp.

Application

For enumeration acc. to the MPN method, and for the selective enrichment of staphylococci from food.
Recommended by the "Schweizerisches Lebensmittelbuch" 5th ed., chapter 56A.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A medium for the isolation of staphylococci from foodstuffs.
G Giolitti et al.
The Journal of applied bacteriology, 29(2), 395-398 (1966-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service