Skip to Content
Merck
All Photos(1)

Key Documents

6.19119

Sigma-Aldrich

N-Chlorosuccinimide

purified grade

Synonym(s):

N-Chlorosuccinimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4ClNO2
CAS Number:
Molecular Weight:
133.53
MDL number:
UNSPSC Code:
12352111

grade

purified grade

SMILES string

ClN1C(=O)CCC1=O

InChI

1S/C4H4ClNO2/c5-6-3(7)1-2-4(6)8/h1-2H2

InChI key

JRNVZBWKYDBUCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service