Skip to Content
Merck
All Photos(1)

Key Documents

XX0045

Supelco

p-Xylene

Reagent

Synonym(s):

p-Xylene, 1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
p-C6H4(CH3)2
CAS Number:
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-396-5
bp:
138.4 °C/1013 hPa
vapor pressure:
8.7 hPa ( 20 °C)

vapor pressure

8.7 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

color

APHA: ≤10

bp

138.4 °C/1013 hPa

mp

13.3 °C

transition temp

flash point 24 °C

solubility

0.2 g/L

density

0.86 g/cm3 at 20  °C

shipped in

ambient

storage temp.

room temp

SMILES string

Cc1ccc(cc1)C

InChI

1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3

InChI key

URLKBWYHVLBVBO-UHFFFAOYSA-N

Application

Reagent

Analysis Note

Assay (GC)98% minColor (APHA)10 maxFormClear liquid (m.p. 13°C)Infrared SpectrumConforms to standard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service