Skip to Content
Merck
All Photos(1)

Key Documents

08086

Millipore

Nitrate Reagent Disks

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

shelf life

limited shelf life, expiry date on the label

application(s)

microbiology

storage temp.

2-8°C

General description

Nitrate Reagent Disks are used to detect an organism′s ability to reduce nitrate. The test involves detection of the enzyme nitrate reductase which causes the reduction of nitrate in the presence of a suitable electron donor to nitrite, which can be tested by an appropriate colorimetric reagent.

Packaging

Sterile filter paper discs (50 pcs, diameter 6 mm) impregnated with Nitrate reagent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service