792934
Specially Denatured Alcohol
190 proof, SDA 2B-4, contains Heptanes
Synonym(s):
Ethanol, SDA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
liquid
contains
Heptanes
concentration
190 proof
application(s)
life science and biopharma
Tax and license information
Permit for use required
SMILES string
OCC
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3
InChI key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
5GA is in metal drum with flex spout.
5GA-PB is in a poly drum with flex spout.
3x5GA is in metal drum with flex spout.
3x5GA-PB is in a poly drum with flex spout.
55GA is in metal drum.
55GA-PB is in poly drum.
5GA-PB is in a poly drum with flex spout.
3x5GA is in metal drum with flex spout.
3x5GA-PB is in a poly drum with flex spout.
55GA is in metal drum.
55GA-PB is in poly drum.
Other Notes
Ordering Information:
To purchase Specially Denatured Alcohol, customers must possess an Industrial Alcohol User Permit from the Alcohol and Tobacco Tax and Trade Bureau (TTB).
TTB Link
To purchase Specially Denatured Alcohol, customers must possess an Industrial Alcohol User Permit from the Alcohol and Tobacco Tax and Trade Bureau (TTB).
- Permit must have customer signature and date of submission to Sigma-Aldrich in addition to the original signature and date.
- Permit must have word, "copy" handwritten or stamped.
- Permit number must contain correct state or US designation matching ship-to address.
- Permit address must match ship-to-address in Sigma-Aldrich system.
- Permit only needs to be provided on first order to sigma-aldrichproductscreening@sial.com.
- All orders require that a PO with the statement, "valid permit #####" be submitted to sigma-aldrichproductscreening@sial.com.
TTB Link
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Soft matter, 11(25), 5053-5059 (2015-06-02)
Cells require adhesion to survive, proliferate and migrate, as well as for wound healing and many other functions. The strength of contractile cell forces on an underlying surface is a highly relevant quantity to measure the affinity of cells to
Toxicology in vitro : an international journal published in association with BIBRA, 29(5), 834-844 (2015-03-31)
In vitro effects of the carbamates pirimicarb and zineb and their formulations Aficida® (50% pirimicarb) and Azzurro® (70% zineb), respectively, were evaluated in Chinese hamster ovary (CHO-K1) cells. Whereas the cytokinesis-blocked micronucleus cytome assay was employed to test for genotoxicity
Chemosphere, 135, 38-45 (2015-04-22)
The occurrence of intersex fish, where male reproductive tissues show evidence of feminization, have been found in freshwater systems around the world, indicating the potential for significant endocrine disruption across species in the ecosystem. Estrogens from birth control medications in
Toxicological sciences : an official journal of the Society of Toxicology, 147(1), 270-285 (2015-06-28)
Traditional toxicology testing has relied on low-throughput, expensive mammalian studies; however, timely testing of the large number of environmental toxicants requires new in vitro and in vivo platforms for inexpensive medium- to high-throughput screening. Herein, we describe the suitability of the asexual
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service