HPLC Analysis of Crufomate (Ruelene) Enantiomers on Astec® CYCLOBOND I 2000
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 μm particles (20024AST)
mobile phase
[A] acetonitrile; [B] acetic acid; [C] triethylamine; (100:0.2:0.1, A:B:C)
flow rate
1 mL/min
column temp.
ambient
detector
UV, 254 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null