6.07375
Lead(II) acetate trihydrate
GR
Synonym(s):
Lead(II) acetate trihydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3COO)2Pb · 3 H2O
CAS Number:
Molecular Weight:
379.33
MDL number:
UNSPSC Code:
12352302
Recommended Products
SMILES string
[Pb](OC(=O)C)OC(=O)C.O.O.O
InChI
1S/2C2H4O2.3H2O.Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);3*1H2;/q;;;;;+2/p-2
InChI key
MCEUZMYFCCOOQO-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service