Skip to Content
Merck
All Photos(1)

Key Documents

6.17604

Supelco

Ethylenediaminetetraacetic acid solution

0.02 N

Synonym(s):

Ethylenediaminetetraacetic acid solution, EDTA solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O8
CAS Number:
Molecular Weight:
292.24
Beilstein:
1716295
MDL number:
UNSPSC Code:
12352116

form

solution

SMILES string

OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)

InChI key

KCXVZYZYPLLWCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application



  • Optimal antigen retrieval for ethanol-fixed cytologic smears.: This study focuses on the optimal antigen retrieval process for ethanol-fixed cytologic smears using EDTA solution. The research highlights the effectiveness of EDTA in improving antigen retrieval, which is crucial for accurate diagnosis and analysis in cytopathology (Denda et al., 2012).


  • A new and reliable culture system for superficial low-grade urothelial carcinoma of the bladder.: This paper describes the development of a new culture system for low-grade urothelial carcinoma using EDTA solution. The research demonstrates that EDTA helps maintain cell viability and integrity, making it a valuable tool in cancer research and diagnosis (Seifert et al., 2007).


Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service