Skip to Content
Merck
All Photos(1)

Key Documents

6.17823

Supelco

Paraffin wax

parameter 60-62 °C

Synonym(s):

Paraffin wax

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12181503

form

solid

parameter

60-62 °C

viscosity

3-6mm2/s(100 °C) (212 °F)

mp

43-95 °C (109 - 203 °F at ca.1,013.25 hPa)

density

0.79-0.94  g/cm3 at 15 °C (59 °F)

SMILES string

CCCNCC(COC1=CC=CC=C1C(=O)CCC2=CC=CC=C2)O

InChI

InChI=1S/C21H27NO3/c1-2-14-22-15-18(23)16-25-21-11-7-6-10-19(21)20(24)13-12-17-8-4-3-5-9-17/h3-11,18,22-23H,2,12-16H2,1H3

InChI key

JWHAUXFOSRPERK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Engineering microporosity in bacterial cellulose scaffolds.: This article details the development of microporous structures within bacterial cellulose scaffolds, potentially for biomedical applications, where paraffin wax might be used as a porogen to enhance the structure′s functional properties (Bäckdahl et al., 2008).

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

602.6 °F - Pensky-Martens closed cup

Flash Point(C)

317 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service