Skip to Content
Merck
All Photos(1)

Key Documents

6.17854

Sigma-Aldrich

Hexane, mixture of isomers

for chromatography

Synonym(s):

Hexanes

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
MDL number:
UNSPSC Code:
41116105

product name

Hexanes, for chromatography

grade

for chromatography

Quality Level

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexanes are a group of hydrocarbon compoundscommonly used in chromatography as a solvent. They are volatile and have lowboiling points, making them suitable for separating and analyzing varioussubstances, including those in the oil industry.

Application

Hexanes can be used to analyze the:

  • Fatty acids derived from lipid A using gas chromatography-mass spectrometry (GC-MS).
  • MPs(membrane proteins) using gas chromatography/mass spectrometry (GC/MS).

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The msbB mutant of Neisseria meningitidis strain NMB has a defect in lipooligosaccharide assembly and transport to the outer membrane
Post DM, et al.
Infection and Immunity, 71(2), 647-655 (2003)
Characterization of htrB and msbB mutants of the light organ symbiont Vibrio fischeri
Adin DM, et al.
Applied and Environmental Microbiology, 74(3), 633-644 (2008)
HPLC/APCI mass spectrometry of saturated and unsaturated hydrocarbons by using hydrocarbon solvents as the APCI reagent and HPLC mobile phase
Gao J, et al.
Journal of the American Society For Mass Spectrometry, 23(5), 816-822 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service