Skip to Content
Merck
All Photos(1)

Key Documents

6.17926

Supelco

Barium sulfate

purified grade

Synonym(s):

Barium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaO4S
CAS Number:
Molecular Weight:
233.39
MDL number:
UNSPSC Code:
12352302
grade:
purified grade

grade

purified grade

SMILES string

[Ba+2].[S](=O)(=O)([O-])[O-]

InChI

1S/Ba.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

TZCXTZWJZNENPQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service