6.18401
Bromocresol green
pH indicator
Synonym(s):
Bromocresol green
About This Item
Recommended Products
Product Name
Bromocresol green, pH 3.6-5.4
pH
3.6-5.4
SMILES string
[S]1(=O)(=O)OC(c4c1cccc4)(c3c(c(c(c(c3)Br)O)Br)C)c2c(c(c(c(c2)Br)O)Br)C
InChI
1S/C21H14Br4O5S/c1-9-12(7-14(22)19(26)17(9)24)21(13-8-15(23)20(27)18(25)10(13)2)11-5-3-4-6-16(11)31(28,29)30-21/h3-8,26-27H,1-2H3
InChI key
FRPHFZCDPYBUAU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Removal of Bromocresol Green from aqueous solution by electro-Fenton and electro-Fenton-like processes with different catalysts: laboratory and kinetic model investigation.: This study explores the efficiency of electro-Fenton and electro-Fenton-like processes in removing Bromocresol Green from aqueous solutions using various catalysts, with a focus on laboratory experimentation and kinetic modeling (Matyszczak et al., 2021).
- Poly (bromocresol green) flakes-decorated pencil graphite electrode for selective electrochemical sensing applications and pharmacokinetic studies.: This research introduces a novel electrochemical sensor using poly(bromocresol green) flakes on pencil graphite electrodes, designed for selective sensing in analytical applications and pharmacokinetic studies (Gadallah et al., 2019).
- Electrocatalytic degradation of bromocresol green wastewater on Ti/SnO(2)-RuO(2) electrode.: This paper examines the degradation of Bromocresol Green in wastewater using an electrocatalytic process on a Ti/SnO2-RuO2 electrode, highlighting its efficiency in pollutant removal (Bai et al., 2017).
- Sensitive and selective spectrophotometric assay of piroxicam in pure form, capsule and human blood serum samples via ion-pair complex formation.: This study presents a spectrophotometric method for assaying piroxicam using Bromocresol Green to form ion-pair complexes, enabling sensitive and selective measurement in various sample types (Alizadeh et al., 2014).
- A high-throughput colorimetric assay to measure the activity of glutamate decarboxylase.: This paper develops a high-throughput colorimetric assay using Bromocresol Green for the rapid measurement of glutamate decarboxylase activity, facilitating enzymatic studies (Yu et al., 2011).
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service