Skip to Content
Merck
All Photos(1)

Key Documents

30483

Sigma-Aldrich

Anti-Mouse IgG−Abberior® CAGE 635 antibody produced in goat

for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)

Synonym(s):

Abberior® CAGE 635-Anti-Mouse-IgG antibody produced in goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Pricing and availability is not currently available.

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

availability

available only in USA and Canada

concentration

~1 mg/mL

fluorescence

λex 630 nm; λem 653 nm in PBS, pH 7.4 (photoactivated)

General description

Abberior CAGE 635 is a caged dye which is initially colorless and nonfluorescent. When illuminated with UV light it undergoes a rapid uncaging reaction and releases a highly fluorescent red dye.
It can most effectively be excited from 620 to 645 nm. Abberior CAGE 635 is designed for single-molecule photoswitching microscopy modes such as PALM, STORM and GSDIM and, in its uncaged form, also performs well in STED microscopy. It performs very well with Leica GSD microscope and other superresolution microscopes such as Nikon N-Storm.
Further, after photoactivation, Abberior CAGE markers can be tracked in confocal and epifluorescence microscopy to analyses molecular dynamics such as diffusion, flow directions and velocities, e.g. are tracking experiments in which the fluorescence signal is observed over time after the initial release of the fluorophore.


Photophysical properties (carboxylic acid):
Absorption Maximum, λex [nm]: 630 (photoactivated, PBS pH 7.4), 316 (non-photoactivated, PBS pH 7.4)
Extinction Coefficient, εmax [M-1cm-1]: 110 000 (photoactivated, PBS pH 7.4)
Photoactivation Wavelength, λ [nm]: 360-440
Fluorescence Maximum, λem [nm]: 653 (photoactivated, PBS pH 7.4)
Recommended STED Wavelength, λ [nm]: 750-800
Fluorescence Quantum Yield, λ: 0,75 (photoactivated, PBS pH 7.4)
Fluorescence Lifetime, τ [ns]: 3,5 (photoactivated, PBS pH 7.4)

Suitability

Designed and tested for fluorescent super-resolution microscopy

Analysis Note

May contain significant amounts of BSA as stabilizer
unconjugated dye ≤5% of total fluorescence

Legal Information

abberior is a registered trademark of Abberior GmbH

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodamines NN: a novel class of caged fluorescent dyes.
Vladimir N Belov et al.
Angewandte Chemie (International ed. in English), 49(20), 3520-3523 (2010-04-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service