Skip to Content
Merck
All Photos(1)

Key Documents

PLA0132

Sigma-Aldrich

Rabbit anti-Sirt1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

SIR2, SIR2-like protein 1, SIR2L1, SIR2alpha, hSIR2, hSIRT1, homolog) 1, regulatory protein SIR2 homolog 1, sirtuin (silent mating type information regulation 2 homolog) 1 (S. cerevisiae), sirtuin (silent mating type information regulation 2; S. cerevisiae, sirtuin type 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunoprecipitation (IP): 1-4 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_036370.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... Sirt1(23411)

Immunogen

The epitope recognized by PLA0132 maps to a region between residue 700 and the C-terminus (residue 747) of human Silent Mating Type Information Regulation 2 homolog 1 using the numbering given in entry NP_036370.2 (GeneID 23411).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Sirt1 is one of seven members of the sirtuin family of proteins that are homologous to the yeast silent information regulator 2 (Sir2), a NAD÷-dependent protein deacetylase that plays a role in lifespan in lower eukaryotes. The sirtuins are diverse in their cellular functions, and are important to the regulation of oxidative stress, and metabolism. Sirt 1 is localized to the nucleus and possesses deacetylase activity. Sirt 1 has been shown to be able to deacetylate p53, Ku70, NF-kappaB and forkhead proteins to mediate cellular resistance to stress.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service