V900442
Choline chloride
Vetec™, reagent grade, ≥98%
Synonym(s):
(2-Hydroxyethyl)trimethylammonium chloride
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
grade
reagent grade
product line
Vetec™
Assay
≥98%
form
solid
color
white
mp
302-305 °C (dec.) (lit.)
SMILES string
[Cl-].C[N+](C)(C)CCO
InChI
1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1
InChI key
SGMZJAMFUVOLNK-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Substrates
Choline acetyltransferase substrate
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Biochimica et biophysica acta, 1828(8), 1856-1862 (2013-04-02)
To better understand the relationship between the relative cytotoxicity of diluted ionic liquids and their specific interaction with biological membranes, the thermotropic behavior of model lipid membrane systems formulated in a series of choline based organic salts was investigated. Unilamellar
Molecular aspects of medicine, 34(2-3), 646-654 (2013-03-20)
The Na(+)-independent, high affinity choline carrier system proposed to supply choline for the synthesis of cell membrane phospholipids was recently associated with SLC44 family members (SLC44A1-5) also called choline-like transporter family. SLC44A1 is widely expressed throughout the nervous system in
European journal of nuclear medicine and molecular imaging, 40(9), 1356-1364 (2013-05-15)
The aims of the study were (a) to evaluate the diagnostic role, by means of positive detection rate (PDR), of ¹⁸F-choline (CH) positron emission tomography (PET)/CT in patients with prostate cancer treated with radiotherapy, with curative intent, and suspicion of
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(6), 833-840 (2013-04-06)
We evaluated the potential of (18)F-fluoromethyldimethyl-2-hydroxyethyl-ammonium (FCH) PET/CT in the detection of recurrent disease or distant metastases and correlated its diagnostic accuracy with prostate-specific antigen (PSA) levels in prostate cancer patients with biochemical evidence of recurrence. Furthermore, the influences of
Nature medicine, 19(5), 576-585 (2013-04-09)
Intestinal microbiota metabolism of choline and phosphatidylcholine produces trimethylamine (TMA), which is further metabolized to a proatherogenic species, trimethylamine-N-oxide (TMAO). We demonstrate here that metabolism by intestinal microbiota of dietary L-carnitine, a trimethylamine abundant in red meat, also produces TMAO
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service