Merck
  • Parallel synthesis of trisubstituted formamidines: a facile and versatile procedure.
Parallel synthesis of trisubstituted formamidines: a facile and versatile procedure.
Journal of combinatorial chemistry (2008-12-05)
Andrea Porcheddu, Giampaolo Giacomelli, Ivana Piredda
ABSTRACT
A focused library of formamidines having several diversity points has been prepared. The use of isocyanate resin as scavenger may offer several practical advantages over conventional procedures, such as the ease of workup and availability. The technique described herein was performed in a parallel synthesis format. The whole protocol has potential use in high throughput synthesis for the preparation and purification of libraries of unsymmetrical formamidines containing sets of different aliphatic substituents.
Sigma-Aldrich
Formamidine acetate salt, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formamidine hydrochloride, 97%