Merck
  • Hypoxia up-regulates SERPINB3 through HIF-2α in human liver cancer cells.
Hypoxia up-regulates SERPINB3 through HIF-2α in human liver cancer cells.
Oncotarget (2014-12-30)
Stefania Cannito, Cristian Turato, Claudia Paternostro, Alessandra Biasiolo, Sebastiano Colombatto, Irene Cambieri, Santina Quarta, Erica Novo, Elisabetta Morello, Gianmarco Villano, Silvano Fasolato, Tiziana Musso, Ezio David, Ignazia Tusa, Elisabetta Rovida, Riccardo Autelli, Antonina Smedile, Umberto Cillo, Patrizia Pontisso, Maurizio Parola
ABSTRACT
SERPINB3 is a cysteine-proteases inhibitor up-regulated in a significant number of cirrhotic patients carrying hepatocellular carcinoma (HCC) and recently proposed as a prognostic marker for HCC early recurrence. SERPINB3 has been reported to stimulate proliferation, inhibit apoptosis and, similar to what reported for hypoxia, to trigger epithelial-to-mesenchymal transition (EMT) and increased invasiveness in liver cancer cells. This study has investigated whether SERPINB3 expression is regulated by hypoxia-related mechanisms in liver cancer cells. Exposure of HepG2 and Huh7 cells to hypoxia up-regulated SERPINB3 transcription, protein synthesis and release in the extracellular medium. Hypoxia-dependent SERPINB3 up-regulation was selective (no change detected for SERPINB4) and operated through hypoxia inducible factor (HIF)-2α (not HIF-1α) binding to SERPINB3 promoter, as confirmed by chromatin immuno-precipitation assay and silencing experiments employing specific siRNAs. HIF-2α-mediated SERPINB3 up-regulation under hypoxic conditions required intracellular generation of ROS. Immuno-histochemistry (IHC) and transcript analysis, performed in human HCC specimens, revealed co-localization of the two proteins in liver cancer cells and the existence of a positive correlation between HIF-2α and SERPINB3 transcript levels, respectively. Hypoxia, through HIF-2α-dependent and redox-sensitive mechanisms, up-regulates the transcription, synthesis and release of SERPINB3, a molecule with a high oncogenic potential.
Sigma-Aldrich
Anti-Goat IgG (whole molecule) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Mouse IgG (whole molecule) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution