BR144515
BRAND® conical joint stopper, borosilicate glass
joint: ST/NS 12/21, solid
Synonym(s):
Conical joint stoppers, Stoppers
All Photos(1)
About This Item
UNSPSC Code:
41122500
NACRES:
NB.51
Recommended Products
material
glass
octagonal borosilicate glass (grip)
Joint
joint: ST/NS12/21
description
octagonal grip
packaging
pack of 1 ea
manufacturer/tradename
BRAND 144515
Related Categories
General description
DIN 12252. Solid or semi-hollow, with octagonal grip.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)