Skip to Content
Merck
All Photos(1)

Key Documents

23370-U

Supelco

Empty Air Monitoring Cassettes

size 37 mm, 3 piece (with spacer ring)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

Pricing and availability is not currently available.

feature

3 piece (with spacer ring)

packaging

pkg of 100 ea

size

37 mm

application(s)

environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

Empty Air Monitoring Cassettes are used for the active sampling of air samples. They are cited in numerous Occupational Health and Safety Administration (OSHA) and National Institute of Occupational Safety and Health (NIOSH) methods in the United States and are universally respected for quality and reliability.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service