Skip to Content
Merck
All Photos(2)

Key Documents

06-946

Sigma-Aldrich

Anti-hyperacetylated Histone H4 (Penta) Antibody

serum, Upstate®

Synonym(s):

H4 histone family, member A, histone 1, H4a, histone cluster 1, H4a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, eukaryotes, Tetrahymena sp.

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Penta-acetylation)

Gene Information

human ... H4C1(8359)

General description

Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Acetylation of histone H4 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs).

Specificity

Expected to cross-react with Tetrahymena and Eukaryote.
Other species not tested.
Recognizes hyperacetylated histone H4, Mr 10 kDa (SDS-PAGE); tri- and tetra-acetylated histone H4 on acid-urea gels. May cross-react with other acetylated proteins.

Immunogen

KLH-conjugated, synthetic peptide
(AGGAcKGGAcKGGAcKGGAcKGGAcKGGC) related to amino acids 2 to 19 of Tetrahymena thermophila histone H4.

Application

Chromatin Immunoprecipitation:
Reported to immunoprecipitate transcriptionally active chromatin containing acetylated histone H4. (Perry, C.A., 1993)

Immunocytochemistry:
Reported to exclusively stain Tetrahymena macronuclei1 and chromatin in HeLa nuclei. (Perry, C.A., 1993)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-hyperacetylated Histone H4 (Penta) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, ICC, WB to detect hyperacetylated Histone H4 (Penta) also known as H4 histone family member A.

Quality

Routinely evaluated by Western Blot on HepG2 lysates.

Western Blot Analysis:
1:1000 dilution of this lot detected HYPERACETYLATED HIS H4 on 10 μg of HepG2 lysates.

Target description

Approx. 10 kDa

Physical form

Rabbit polyclonal antiserum IgG in buffer containing 30% glycerol.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variabillity in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Tetrahymena cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The PB2 subunit of the influenza virus RNA polymerase affects virulence by interacting with MAVS and inhibiting IFN-{beta} expression.
Graef KM, Vreede FT, Lau YF, McCall AW, Carr SM, Subbarao K, Fodor E
Journal of virology null
A network connecting Runx2, SATB2, and the miR-23a~27a~24-2 cluster regulates the osteoblast differentiation program.
Hassan, MQ; Gordon, JA; Beloti, MM; Croce, CM; van Wijnen, AJ; Stein, JL; Stein, GS; Lian, JB
Proceedings of the National Academy of Sciences of the USA null
Cindy Law et al.
The Journal of biological chemistry, 290(21), 13234-13249 (2015-04-04)
H2A.Z is a histone H2A variant that is essential for viability in Tetrahymena and Drosophila and also during embryonic development of mice. Although implicated in diverse cellular processes, including transcriptional regulation, chromosome segregation, and heterochromatin formation, its essential function in
Manjeet Kaur Gill-Sharma et al.
Clinical epigenetics, 4(1), 23-23 (2012-12-18)
The putative regulatory role of the male reproductive hormones in the molecular mechanism underlying chromatin condensation remains poorly understood. In the past decade, we developed two adult male rat models wherein functional deficits of testosterone or FSH, produced after treatments
Leslie Mitchell et al.
PloS one, 6(10), e25336-e25336 (2011-10-11)
In the budding yeast Saccharomyces cerevisiae, the lysine acetyltransferase NuA4 has been linked to a host of cellular processes through the acetylation of histone and non-histone targets. To discover proteins regulated by NuA4-dependent acetylation, we performed genome-wide synthetic dosage lethal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service