Skip to Content
Merck
All Photos(1)

Key Documents

07-1585

Sigma-Aldrich

Anti-HIF-1-alpha Antibody, hydroxyproline (Pro402)

from rabbit, purified by affinity chromatography

Synonym(s):

Hypoxia-inducible factor 1-alpha, HIF-1-alpha, HIF1-alpha, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, Class E basic helix-loop-helix protein 78, bHLHe78, Member of PAS protein 1, PAS domain-containing protein 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), zebrafish (based on 100% sequence homology), mouse (based on 100% sequence homology), bovine (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... HIF1A(3091)

General description

Cell growth and viability is compromised by oxygen deprivation (hypoxia). Hypoxia-inducible factors, including HIF-1α, HIF-1β (also designated Arnt 1), EPAS-1 (also designated HIF-2α) and HIF-3α, induce glycolysis, erythropoiesis and angiogenesis in order to restore oxygen homeostasis. Hypoxia-inducible factors are members of the Per-Arnt-Sim (PAS) domain transcription factor family. In response to hypoxia, HIF-1α is upregulated and forms a heterodimer with HIF-1β to form the HIF-1 complex. The HIF-1 complex recognizes and binds to the hypoxia responsive element (HRE) of hypoxia-inducible genes, thereby activating transcription. Hypoxia-inducible expression of some genes such as Glut-1, p53, p21 or Bcl-2, is HIF-1α dependent, whereas expression of others, such as p27, GADD 153 or HO-1, is HIF-1α independent. EPAS-1 and HIF-3α have also been shown to form heterodimeric complexes with Arnt 1 in response to hypoxia.

Specificity

This antibody recognizes HIF-1-alpha at the ODD region.

Immunogen

Epitope: ODD region
KLH-conjugated linear peptide corresponding to the ODD region of human HIF-1-alpha.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-HIF-1-alpha Antibody, hydroxyproline (Pro402) (Rabbit Polyclonal Antibody) validated in WB to detect HIF-1-alpha also known as Hypoxia-inducible factor 1-alpha, HIF1-alpha.

Quality

Evaluated by Western Blot in FeCl2 treated HIF-1-alpha recombinant proteins.

Western Blot Analysis: 1 µg/mL of this antibody detected HIF-1-alpha on 10 µg of FeCl2 treated HIF-1-alpha recombinant proteins.

Target description

~120 kDa observed using Gal4-tagged recombinant protein. 92 kDa calculated for native protein.

Linkage

Replaces: 04-1006

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
FeCl2 treated HIF-1-alpha recombinant proteins

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael I Koukourakis et al.
Scientific reports, 6, 30986-30986 (2016-08-11)
The mechanism of Amifostine (WR-2721) mediated radioprotection is poorly understood. The effects of amifostine on human basal metabolism, mouse liver metabolism and on normal and tumor hepatic cells were studied. Indirect calorimetric canopy tests showed significant reductions in oxygen consumption
Ya-Min Tian et al.
The Journal of biological chemistry, 286(15), 13041-13051 (2011-02-22)
Hypoxia inducible factor (HIF) is regulated by dual pathways involving oxygen-dependent prolyl and asparaginyl hydroxylation of its α-subunits. Prolyl hydroxylation at two sites within a central degradation domain promotes association of HIF-α with the von Hippel-Lindau ubiquitin E3 ligase and
Linda O'Flaherty et al.
Human molecular genetics, 19(19), 3844-3851 (2010-07-28)
Mutations in the gene encoding the Krebs cycle enzyme fumarate hydratase (FH) predispose to hereditary leiomyomatosis and renal cell cancer in affected individuals. FH-associated neoplasia is characterized by defective mitochondrial function and by upregulation of transcriptional pathways mediated by hypoxia-inducible
A Lakoma et al.
Cell death discovery, 1 (2015-01-01)
Neuroblastoma is an aggressive pediatric malignancy which is >98% p53 wild-type at diagnosis. As a primary repressor of p53 activity and part of a p53-activated negative feedback loop, targeting of mouse double minute 2 homolog (MDM2) is an attractive therapeutic
Proline-hydroxylated hypoxia-inducible factor 1? (HIF-1?) upregulation in human tumours.
Snell, CE; Turley, H; McIntyre, A; Li, D; Masiero, M; Schofield, CJ; Gatter, KC; Harris et al.
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service