Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

1.08310

Supelco

o-Toluidine

GR for analysis

Synonym(s):

o-Toluidine, 2-Aminotoluene, 2-Methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(CH3)C6H4NH2
CAS Number:
Molecular Weight:
107.15
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-429-0

vapor pressure

0.18 hPa ( 20 °C)

Quality Level

Assay

≥99.5% (GC)

autoignition temp.

480 °C

potency

3250 mg/kg LD50, skin (Rabbit)

expl. lim.

1.5-7.5 % (v/v)

pH

7.4 (20 °C in H2O, Aqueous solution)

bp

200 °C/1013 hPa

mp

<-15 °C

transition temp

flash point 85 °C

solubility

15 g/L

density

0.99 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5H,8H2,1H3

InChI key

RNVCVTLRINQCPJ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.5 % (a/a)
p-toluidine and m-toluidine (GC, area%): ≤ 0.40 % (a/a)
Water (K. F.): ≤ 0.10 %
Chloride (Cl): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0001 %
Sulfated ash: ≤ 0.005 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

PRTR

NON

FSL

Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service