Skip to Content
Merck
All Photos(2)

Key Documents

1.10663

Millipore

Test agar pH 6.0 for the inhibitor test

for microbiology

Synonym(s):

Standard nutrient agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

Quality Level

form

powder (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.5 (37 °C, 25 g/L in H2O, after autoclaving)

solubility

25 g/L

bulk density

670 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish-brown
pH-value (25 °C): 5.9 - 6.1
Solidification behaviour (2 hrs., 40 °C): liquid
Inhibition zone diameter/Gentamicin 10 µg (Bacillus subtilis BGA): 20 - 28 mm
Inhibition zone diameter/Gentamycin 30 µg (Bacillus subtilis BGA): 22 - 30 mm
Inhibition zone diameter/Penicillin G 10 U/IE (Bacillus subtilis BGA): 36 - 48 mm
Inhibition zone diameter/Streptomycin 10 µg (Bacillus subtilis BGA): 19 - 27 mm

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service