208711
Ca2+/Calmodulin Kinase II Inhibitor 281-309
The Ca2+/Calmodulin Kinase II Inhibitor 281-309 controls the biological activity of Ca2+/Calmodulin Kinase II. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
Ca2+/Calmodulin Kinase II Inhibitor 281-309, CaM Kinase II Inhibitor 281-309, MHRQETVDCLKKFNARRKLKGAILTTMLA-OH
About This Item
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
lyophilized solid
potency
80 nM IC50
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
solubility
water: 5 mg/mL
shipped in
ambient
storage temp.
−20°C
General description
Biochem/physiol Actions
Packaging
Warning
Sequence
Other Notes
Waxham, M.N., et al. 1993. Brain Res. 609, 1.
Fukunaga, K., et al. 1990. J. Neurochem.54, 103.
Colbran, R.J., et al. 1989. J. Biol. Chem. 264, 4800.
Colbran, R.J., et al. 1988. J. Biol. Chem.263, 18145.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
208711-500UG:
208711-UG:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service