Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

496265

Sigma-Aldrich

Oncrasin-1

A cell-permeable indolecarboxaldehyde compound that selectively induces apoptosis in cells harboring mutant K-Ras both in vitro and in vivo.

Synonym(s):

Oncrasin-1, 1-((4-Chlorobenzyl)-1H-indole-3-carboxaldehyde, Oncogenic Ras Tumor- Inhibiting compound 1, 1-((4-Chlorobenzyl)-1H-indole-3-carboxaldehyde, Oncogenic Ras Tumor-Inhibiting compound 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12ClNO
CAS Number:
Molecular Weight:
269.73
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white to light pink

solubility

DMSO: soluble
ethanol: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C16H12ClNO/c17-14-7-5-12(6-8-14)9-18-10-13(11-19)15-3-1-2-4-16(15)18/h1-8,10-11H,9H2

InChI key

ZDRQMXCSSAPUMM-UHFFFAOYSA-N

General description

A cell-permeable indolecarboxaldehyde compound that selectively induces apoptosis in cells harboring mutant K-Ras both in vitro (IC50 = 0.11, 0.25, 1.0, 1.58, and 4.81 µM in H2887, H460, H2122, A549, and T29Kt1 cultures, respectively) and in vivo (75% suppression of H460-derived tumor growth in mice; 100 mg/kg via daily i.p.), while exhibiting little or much reduced potency toward cells with wild-type Ras or mutant N-Ras (IC50 ≥31 µM). Oncrasin-1 treatment is shown to induce PKCι aggregation with splicing factors in the nucleus and dissociation between PKCι and CDK9/Cyclin T1 in K-Ras mutant cultures, resulting in decreased RNA polymerase II phosphorylation and a suppression of RNA processing in general. Oncrasin-1-induced cell death requires the presence of both mutant K-Ras and PKCι, siRNA-mediated knockdown of either enzyme is demonstrated to abolish its cytotoxicity in H460 and T29Kt1 cultures.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Guo, W., et al. 2009. Mol. Cancer Ther.8, 441.
Guo, W., et al. 2008. Cancer Res.68, 7403.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

496265-MG:
496265-25MG:
496265-1.1ML:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service