Skip to Content
Merck
All Photos(1)

Key Documents

5.08505

Sigma-Aldrich

A-803467

Synonym(s):

A-803467, Sodium Channel Type 1.8 (Nav1.8) blocker, A-803467, A 803467, A803467

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16ClNO4
CAS Number:
Molecular Weight:
357.79
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

8 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mM
ethanol: 25 mM

storage temp.

2-8°C

InChI

1S/C19H16ClNO4/c1-23-15-9-14(10-16(11-15)24-2)21-19(22)18-8-7-17(25-18)12-3-5-13(20)6-4-12/h3-11H,1-2H3,(H,21,22)

InChI key

VHKBTPQDHDSBSP-UHFFFAOYSA-N

General description

A voltage gated sodium channel type 1.8 (Nav1.8) pore blocker. Potently blocks tetrodotoxin-resistant currents (IC50) = 140 nM) and the generation of spontaneous and electrically evoked action potentials in vitro in rat dorsal root ganglion neurons. In recombinant cell lines, A-803467 potently blocked human Nav1.8 (IC50 = 8 nM) and was >100-fold selective vs. human Nav1.2, Nav1.3, Nav1.5, and Nav1.7 (IC50 values ≥ 1 µM).

Biochem/physiol Actions

Primary Target
Nav1.8

Warning

Toxicity: Standard Handling (A)

Other Notes

Jarvis, M. F. et al. 2007. Proc. Natl. Acad. Sci. USA104, 8520.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service