Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

565730

Sigma-Aldrich

Scriptaid

A relatively non-toxic, cell-permeable hydroxamic acid-containing histone deacetylase (HDAC) inhibitor.

Synonym(s):

Scriptaid, 6-(1,3-Dioxo-1 H,3 H-benzo[ de]isoquinolin-2-yl)-hexanoic Acid Hydroxyamide, CGK1026, HDAC Inhibitor XIII, 6-(1,3-Dioxo-1H,3H-benzo[de]isoquinolin-2-yl)-hexanoic Acid Hydroxyamide, CGK1026, HDAC Inhibitor XIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N2O4
CAS Number:
Molecular Weight:
326.35
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 4 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C18H18N2O4/c21-15(19-24)10-2-1-3-11-20-17(22)13-8-4-6-12-7-5-9-14(16(12)13)18(20)23/h4-9,24H,1-3,10-11H2,(H,19,21)

InChI key

JTDYUFSDZATMKU-UHFFFAOYSA-N

General description

A relatively non-toxic hydroxamic acid-containing histone deacetylase (HDAC) inhibitor. Facilitates transcriptional activation (TGFβ/Smad4) in both stable and transient reporter assays in a concentration-dependent manner. Useful agent for transfection assays in reporter systems due to the lack of interference with the ability of reporter construct to measure positive activation of transcription factor in response to signal transduction stimulus. It causes over 100-fold increase in histone acetylation in PANC-1 cells at 2 µg/ml (6 µM). Reported to derepress hTERT by inhibiting the recruitment of HDAC into E2F-pocket protein complexes assembled on the hTERT promoter.
A relatively non-toxic, cell-permeable hydroxamic acid-containing histone deacetylase (HDAC) inhibitor. Facilitates transcriptional activation (TGFβ/Smad4) in both stable and transient receptor assays in a concentration-dependent manner. At ~2 µg/ml (6-8 µM) concentrations, results in a greater than 100-fold increase in histone acetylation in PANC-1 cells. Reported to derepress hTERT by inhibiting the recruitment of HDAC into E2F-pocket protein complexes assembled on the hTERT promoter.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
HDAC
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Won, J., et al. 2004. Proc. Natl. Acad. Sci. USA101, 11328.
Su, G.H., et al. 2000. Cancer Res.60, 3137.

Legal Information

Sold under license of U.S. Patent 6,544,957.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

565730-1.1ML:
565730-5MG:
565730-MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service