Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

573105

Sigma-Aldrich

STAT3 Inhibitor Peptide, Cell-permeable, Inactive Control

The STAT3 Inhibitor Peptide, Cell-permeable, Inactive Control controls the biological activity of STAT3.

Synonym(s):

STAT3 Inhibitor Peptide, Cell-permeable, Inactive Control

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C92H156N20O21
Molecular Weight:
1878.34
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable, non-phosphorylated inactive control peptide for STAT3 cellular function studies utilizing STAT3 Inhibitor PpYLKTK-mts (Cat. No. 573096).
The STAT3 Inhibitor Peptide, Cell-permeable, Inactive Control controls the biological activity of STAT3.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Pro-Tyr-Leu-Lys-Thr-Lys-Ala-Ala-Val-Leu-Leu-Pro-Val-Leu-Leu-Ala-Ala-Pro-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Turkson, J., et al. 2001. J. Biol. Chem.276, 45443.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

573105-MG:
573105-5MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service