616385
α-Toxin, Staphylococcus aureus
Alpha Toxin, CAS 12616-52-3, is a major cytotoxin isolated and purified from the Wood 46 strain of Staphylococcus aureus. At low concentration it binds to cell surface receptors and forms heptameric pores.
Synonym(s):
α-Toxin, Staphylococcus aureus
About This Item
Recommended Products
Quality Level
form
solid
specific activity
≥30 units/μg protein
manufacturer/tradename
Calbiochem®
availability
not available in Canada,
storage condition
OK to freeze
storage temp.
−20°C
General description
Biochem/physiol Actions
Stimulates prostaglandin synthesis and phosphorylation of myelin basic protein.
Warning
Unit Definition
Physical form
Preparation Note
Reconstitution
Analysis Note
Other Notes
Cescatti, L., et al. 1991. J. Membr. Biol.119, 53.
Harshman. S., et al. 1988. Methods Enzymol.165, 3.
Hohman, R.J. 1988. Proc. Natl. Acad. Sci. USA 85, 1624.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Inhalation - Acute Tox. 1 Oral
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
616385-UG:
616385-250UG:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service