Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

8.01335

Sigma-Aldrich

2-Aminothiophenol

for synthesis

Synonym(s):

2-Aminothiophenol, 2-Mercaptoaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(NH2)C6H4SH
CAS Number:
Molecular Weight:
125.19
MDL number:
UNSPSC Code:
12352105
EC Index Number:
205-277-3
form:
liquid
Assay:
≥98.0% (GC)

vapor pressure

1 hPa ( 69 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

234 °C/1013 hPa

mp

26 °C

transition temp

flash point 79 °C

solubility

0.027 g/L

density

1.17 g/cm3 at 20 °C

InChI

1S/C6H7NS.ClH/c7-5-3-1-2-4-6(5)8;/h1-4,8H,7H2;1H

InChI key

PEDOSWZBWWCOAY-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.173 - 1.177
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service