Skip to Content
Merck
All Photos(1)

Key Documents

8.14974

Sigma-Aldrich

Phenyllithium

(20% solution in dibutylether) for synthesis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
form:
liquid

Quality Level

form

liquid

transition temp

flash point 25 °C (Information refers to the main component.)

density

0.85 g/cm3 at 20 °C

storage temp.

2-8°C

Analysis Note

Assay (total alcalinity): 17.5 - 20.5 %

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1A - Eye Dam. 1 - Flam. Liq. 3 - Muta. 1B - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Blood, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(C)

25 °C - Information refers to the main component.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service