Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

8.41749

Sigma-Aldrich

1-Pentanethiol

for synthesis

Synonym(s):

1-Pentanethiol, Amyl mercaptan, Pentyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12S
CAS Number:
Molecular Weight:
104.21
MDL number:
UNSPSC Code:
12352105
EC Index Number:
203-789-1

vapor pressure

18.5 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

125-126 °C/1013 hPa

mp

-76 °C

transition temp

flash point 18 °C

solubility

0.16 g/L

density

0.841 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H12S/c1-2-3-4-5-6/h6H,2-5H2,1H3

InChI key

ZRKMQKLGEQPLNS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.840 - 0.844
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Flammable liquids
Type 1 petroleums
Hazardous rank II
Water insoluble liquid


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service