Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

AB3738-I

Sigma-Aldrich

Anti-Beta Lactamase

from rabbit, purified by affinity chromatography

Synonym(s):

Beta-lactamase TEM, IRT-4, Penicillinase, TEM-1, TEM-16/CAZ-7, TEM-2, TEM-24/CAZ-6, TEM-3, TEM-4, TEM-5, TEM-6, TEM-8/CAZ-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
E. coli
technique(s):
western blot: suitable
citations:
1

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

E. coli

species reactivity (predicted by homology)

all

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

General description

Beta-lactamase TEM (EC 3.5.2.6; UniProt P62593; also known as IRT-4, Penicillinase, TEM-1, TEM-16/CAZ-7, TEM-2, TEM-24/CAZ-6, TEM-3, TEM-4, TEM-5, TEM-6, TEM-8/CAZ-2) is encoded by the bla (also known as blaT-3, blaT-4, blaT-5, blaT-6) gene (Gene ID 3244915) in E. coli. TEM-1 belongs to the class-A beta-lactamase family. Beta-lactamases produced by bacteria provide resistance to beta-lactam antibiotics, such as penicillins and cephamycins by breaking open the beta-lactam ring. Over 200 beta-lactamases have been reported that are classified into four main groups (A, B, C, and D) and eight subgroups according to their functional and structural characteristics. The TEM-1 is predominant plasmid-mediated beta-lactamase of gram-negative rods. Six different nucleotide sequences, called blaTEM-1a, blaTEM-1b, blaTEM-1c, blaTEM-1d, blaTEM-1e, and blaTEM-1f, have been described to codify the same TEM-1 beta-lactamase. Ref.: Briñas, L., et al. (2002). Antimicrob. Agents Chemother. 46, 103156-103163.

Specificity

This polyclonal antibody detects E. coli beta-lactamase.

Immunogen

GST-tagged full-length E. coli beta-lactamase, including the signal peptide sequence.

Application

Detect beta-lactamase using this rabbit Anti-Beta Lactamase polyclonal antibody, Cat. No. AB3738-I, validated for use in Western Blotting.

Quality

Evaluated by Western Blotting in lasate from beta-lactamase-transformed E.coli.

Western Blotting Analysis: A 1:100,000 dilution of this antibody detected GST-tagged full-length E. coli beta-lactamase in 0.5 µg of lysate from transformed E. coli.

Target description

31.52/28.91 kDa (with/without signal peptide a.a. 1-23) calculated.

Linkage

Replaces: AB3738

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

AB3738-I:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nils Peiter et al.
Life science alliance, 6(1) (2022-11-01)
Cotranslational insertion of selenocysteine (Sec) proceeds by recoding UGA to a sense codon. This recoding is governed by the Sec insertion sequence (SECIS) element, an RNA structure on the mRNA, but size, location, structure determinants, and mechanism differ for Bacteria

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service