Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

MAB1964

Sigma-Aldrich

Anti-Integrin β4 Antibody, clone 3E1

ascites fluid, clone 3E1, Chemicon®

Synonym(s):

CD104

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
3E1, monoclonal
application:
FACS
ICC
IHC
IP
species reactivity:
human
technique(s):
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
citations:
37

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3E1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ITGB4(3691)

General description

Integrin beta 4 is a glycoprotein which associates with the a6 integrin to form the a6/b4 complex. Integrin alpha 6/beta 4 is a receptor for laminin. It plays a critical structural role in the hemidesmosome of epithelial cells. Defects in Integrin beta 4 gene are a cause of epidermolysis bullosa letalis with pyloric atresia (EB PA); also known as junctional epidermolysis bullosa with pyloric atresia (PA-JEB) or aplasia cutis congenita with gastrointestinal atresia. EB-PA is characterized by mucocutaneous fragility and gastrointestinal atresia, which most commonly affects the pylorus. Moreover, defects in Integrin beta 4 gene are a cause of generalized atrophic benign epidermolysis bullosa (GABEB). This nonlethal form of junctional epidermolysis bullosa is characterized by life long blistering of the skin, associated with hair and tooth abnormalities.

Specificity

Reacts with Human beta4 integrin. The antigen is present on epithelial cells.

Application

Stains human skin in immunofluorescence at approximately 1:200; acetone fixation required.

Also suitable for use in immunoprecipitation and flow cytometric assays.

Optimal working dilutions must be determined by the end user.
This Anti-Integrin β4 Antibody, clone 3E1 is validated for use in FC, IC, IH, IP for the detection of Integrin β4.

Physical form

UnPurified ascites containing no preservatives.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

MAB1964:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Z Gilcrease et al.
Cancer research, 64(20), 7395-7398 (2004-10-20)
Clustering of cell-surface integrins is known to augment integrin-mediated signal transduction, but mechanisms of integrin clustering are poorly understood. Here we report that adhesion-independent clustering of alpha6beta4 integrin, known to be important in mediating tumor cell motility, is driven by
Sho Hiroyasu et al.
The American journal of pathology, 182(3), 828-840 (2013-01-23)
Bullous pemphigoid (BP) is an autoimmune blistering skin disease induced by pathogenic autoantibodies against a type II transmembrane protein (BP180, collagen type XVII, or BPAG2). In animal models, BP180 autoantibody-antigen interaction appears insufficient to develop blisters, but involvement of complement
Rachel M Levine et al.
Bioengineering & translational medicine, 1(2), 168-180 (2016-08-08)
The greatest obstacle to clinical application of cancer gene therapy is lack of effective delivery tools. Gene delivery vehicles must protect against degradation, avoid immunogenic effects and prevent off target delivery which can cause harmful side effects. PEGylated liposomes have
A G Lowrie et al.
British journal of cancer, 91(7), 1327-1334 (2004-09-09)
Interactions between tumour cells and the microenvironment are increasingly recognised to have an influence on cancer progression. In pancreatic carcinoma, a highly desmoplastic stroma with abnormal extracellular matrix (ECM) protein and interleukin-8 (IL-8) expression is seen. To investigate whether the
Emilie Faure et al.
Journal of cell science, 125(Pt 18), 4264-4277 (2012-06-22)
α6β4 integrin is the main component of hemidesmosomes (HD) that stably anchor the epithelium to the underlying basement membrane. Epithelial cell migration requires HD remodelling, which can be promoted by epidermal growth factor (EGF). We previously showed that extracellular nucleotides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service