Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

01-2030

Sigma-Aldrich

Bovine Serum Albumin

SAJ special grade, powder

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202

grade

SAJ special grade

form

powder

mol wt

~66 kDa

composition

N, 14.5-16.4%

availability

available only in Japan

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Preparation Note

Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

01-2030-1-1G-J:
01-2030-2-25G-J:
01-2030-3-5G-J:
01-2030-4-10G-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcos V A S Navarro et al.
PLoS biology, 9(2), e1000588-e1000588 (2011-02-10)
The bacterial second messenger bis-(3'-5') cyclic dimeric guanosine monophosphate (c-di-GMP) has emerged as a central regulator for biofilm formation. Increased cellular c-di-GMP levels lead to stable cell attachment, which in Pseudomonas fluorescens requires the transmembrane receptor LapD. LapD exhibits a
Manoj K Pandey et al.
Molecular pharmacology, 75(3), 525-533 (2008-12-24)
The aim of the current study is to determine whether butein (3,4,2',4'-tetrahydroxychalcone) exhibits antiproliferative effects against tumor cells through suppression of the signal transducer and activator of transcription 3 (STAT3) activation pathway. We investigated the effects of butein on constitutive
Balázs Rada et al.
Journal of immunology (Baltimore, Md. : 1950), 181(7), 4883-4893 (2008-09-20)
The dual oxidase-thiocyanate-lactoperoxidase (Duox/SCN(-)/LPO) system generates the microbicidal oxidant hypothiocyanite in the airway surface liquid by using LPO, thiocyanate, and Duox-derived hydrogen peroxide released from the apical surface of the airway epithelium. This system is effective against several microorganisms that
Robert W Crawford et al.
Infection and immunity, 76(11), 5341-5349 (2008-09-17)
Salmonella enterica serovar Typhi can establish a chronic, asymptomatic infection of the human gallbladder, suggesting that this bacterium utilizes novel mechanisms to mediate enhanced colonization and persistence in a bile-rich environment. Gallstones are one of the most important risk factors
Eva P Browne et al.
Aquatic toxicology (Amsterdam, Netherlands), 92(4), 281-287 (2009-04-07)
Polybrominated diphenyl ethers (PBDEs) are brominated flame retardants that persist in the environment and are present in geographically widespread fish species. PBDE concentrations can be particularly high in resident Chinook salmon (Onchorhynchus tshawytscha) in the Puget Sound, Washington. Although PBDE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service