Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

01-4600

Sigma-Aldrich

Ammonium citrate dibasic

suitable for atomic absorption spectrometry, 98.0-102.0%

Synonym(s):

Ammonium hydrogencitrate, Citric acid ammonium salt, Diammonium hydrogen citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(CO2H)(CH2CO2NH4)2
CAS Number:
Molecular Weight:
226.18
Beilstein:
4925760
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

1.8 (vs air)

Assay

98.0-102.0%

form

solid

availability

available only in Japan

pH

5.2 (20 °C, 50 g/L)

suitability

suitable for atomic absorption spectrometry

SMILES string

N.N.OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7.2H3N/c7-3(8)1-6(13,5(11)12)2-4(9)10;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);2*1H3

InChI key

YXVFQADLFFNVDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

01-4600-5-500G-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorina Gjonaj et al.
Organic & biomolecular chemistry, 13(21), 6059-6065 (2015-05-08)
A new method for the selective chemical modification of DNA at cytosine nucleobases using alkoxy- and benzyloxyamines is presented. It is shown that in particular benzyloxyamines are effective DNA modifying agents, giving rise to almost exclusive formation of the mono
Shi-Jun Dong et al.
The international journal of biochemistry & cell biology, 68, 33-41 (2015-08-19)
During the industrial bioethanol fermentation, Saccharomyces cerevisiae cells are often stressed by bacterial contaminants, especially lactic acid bacteria. Generally, lactic acid bacteria contamination can inhibit S. cerevisiae cell growth through secreting lactic acid and competing with yeast cells for micronutrients
S Vollmer et al.
Organic & biomolecular chemistry, 13(20), 5734-5742 (2015-04-23)
Triplexes with a gap in the purine strand have been shown to bind adenosine or guanosine derivatives through a combination of Watson-Crick and Hoogsteen base pairing. Rigidifying the binding site should be advantageous for affinity. Here we report that clamps
Ewa Radzikowska et al.
Organic & biomolecular chemistry, 13(1), 269-276 (2014-11-05)
Chimeric oligonucleotides containing phosphodiester and phosphorothioate linkages have been obtained using the solid phase synthesis. The oligonucleotide parts possessing natural internucleotide phosphate bonds were assembled using commercially available nucleoside 3'-O-(2-cyanoethyl-N,N-diisopropylamino)phosphoramidites 7 whereas the phosphorothioate segment was built using nucleoside 3'-O-(2-thio-1,3,2-oxathiaphospholanes)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service