Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

05-1050

Sigma-Aldrich

Calcium standard solution

suitable for atomic absorption spectrometry, 1000 ppm Ca+2 in 1 M HCl, 1 mg/mL Ca+2 in 1 M HCl

Synonym(s):

Calcium chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2
CAS Number:
Molecular Weight:
110.98
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

liquid

availability

available only in Japan

concentration

1 mg/mL Ca+2 in 1 M HCl
1000 ppm Ca+2 in 1 M HCl

suitability

suitable for atomic absorption spectrometry

SMILES string

Cl[Ca]Cl

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

05-1050-6-100ML-J:4.548173002941E12


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahisa Kuga et al.
Scientific reports, 10(1), 7621-7621 (2020-05-08)
There are eight human Src-family tyrosine kinases (SFKs). SFK members c-Src, c-Yes, Fyn, and Lyn are expressed in various cancer cells. SFK kinase activity is negatively regulated by Csk tyrosine kinase. Reduced activity of Csk causes aberrant activation of SFKs
Marcos Francisco Núñez et al.
Molecular biology of the cell, 31(7), 667-682 (2019-12-27)
B-cells become activated by ligands with varying valency and mode of presentation to the B-cell receptor (BCR). We previously demonstrated that clustering the immunoglobulin M (IgM) isotype of BCR with an artificial soluble cross-linker stabilized an ordered phase-like domain that
Argiris Papathanasopoulos et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 29(9), 1327-1335 (2011-03-25)
An unexpected side effect of some classes of anticoagulants has been osteoporosis which may be, at least in part, related to deranged mesenchymal stem cell (MSC) function. The aim of the present study was to compare the effect of fondaparinux
Avinash A Kadam et al.
Bioresource technology, 132, 276-284 (2013-02-16)
Pretreatments to sugarcane bagasse (SCB) such as CaCl2, alkali, ammonia, steam and milling showed 91%, 46%, 47%, 42% and 56% adsorption of Solvent Red 5B (SR5B); 92%, 57%, 58%, 56% and 68% adsorption of simulated dyes mixture (SDM), and 86%
Chao-Han Lai et al.
Annals of surgery, 258(6), 1103-1110 (2013-01-09)
To investigate whether recombinant thrombomodulin containing all the extracellular domains (rTMD123) has therapeutic potential against aneurysm development. The pathogenesis of abdominal aortic aneurysm (AAA) is characterized by chronic inflammation and proteolytic degradation of extracellular matrix. Thrombomodulin, a transmembrane glycoprotein, exerts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service