Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

24-4340

Sigma-Aldrich

Potassium chromate

JIS special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CrO4
CAS Number:
Molecular Weight:
194.19
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥99.0%
grade:
JIS special grade
form:
solid

grade

JIS special grade

vapor density

6.7 (vs air)

Assay

≥99.0%

form

solid

availability

available only in Japan

pH

8.5-10.0 (20 °C, 50 g/L)

mp

971 °C (lit.)

SMILES string

[K+].[K+].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.2K.4O/q;2*+1;;;2*-1

InChI key

XMXNVYPJWBTAHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

PRTR

Specified Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

24-4340-2-25G-J:
24-4340-5-500G-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subhendra N Mattagajasingh et al.
Journal of applied toxicology : JAT, 28(8), 987-997 (2008-07-11)
Hexavalent chromium [Cr(VI)] compounds are Group-I human carcinogens. Cr(VI)-induced DNA-protein crosslinks (DPCs) have been implicated in the mutagenic and carcinogenic effects of Cr(VI). Although multiple mechanisms have been suggested for Cr(VI)-induced DNA-protein crosslinking, the mechanism of formation of DNA-protein crosslinks
Xue Zhou et al.
Toxicology and applied pharmacology, 236(1), 78-84 (2009-04-18)
Occupational exposure to nickel (Ni), chromium (Cr), and arsenic (As) containing compounds has been associated with lung cancer and other adverse health effects. Their carcinogenic properties may be attributable in part, to activation and/or repression of gene expression induced by
Cecylia Wardak
Journal of hazardous materials, 186(2-3), 1131-1135 (2010-12-21)
A new polyvinylchloride membrane sensor for Pb(2+) with solid contact based on ionic liquid has been prepared. The electrode shows a Nernstian response for lead ions over a wide concentration range (1×10(-8) to 1×10(-1) mol L(-1)) and the slope of
Saturnino Spiga et al.
Brain structure & function, 216(3), 171-182 (2011-04-05)
Visualization of neuronal elements is of fundamental importance in modern neuroscience. Golgi-Cox impregnation is a widely employed method that provides detailed information about morphological characteristics of neurons, but none regarding their neurochemical features. Immunocytochemical procedures, on the other hand, can
Inês Domingues et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 152(3), 338-345 (2010-06-23)
The present work aims to compare the sensitivity of embryos and adult zebrafish to chromium (VI) (as potassium dichromate) focusing on biomarkers (cholinesterase, glutathione S-transferase and lactate dehydrogenase) as endpoints. Zebrafish eggs showed less sensitivity to Cr (VI) (96 h-LC50=145.7

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service