Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

28-0966

Sigma-Aldrich

Siliceous sand

CP

Synonym(s):

Sand, white quartz, Quartz, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

CP

form

solid

availability

available only in Japan

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

28-0966-5-500G-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Afra et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(4), 045006-045006 (2012-12-15)
The morphology of swift heavy ion tracks in crystalline α-quartz was investigated using small angle x-ray scattering (SAXS), molecular dynamics (MD) simulations and transmission electron microscopy. Tracks were generated by irradiation with heavy ions with energies between 27 MeV and
Jenia Gutman et al.
Environmental science & technology, 47(1), 398-404 (2012-11-29)
This investigation is focused on the combined effect of bacterial physicochemical characteristics and motility on cell adhesion and deposition using a flow-through quartz crystal microbalance with dissipation (QCM-D). Three model flagellated strains with different degrees of motility were selected, including
Yinying Lu et al.
Environmental pollution (Barking, Essex : 1987), 182, 269-277 (2013-08-21)
The effect of different surfactants on the transport of multiwalled carbon nanotubes (MWCNTs) in quartz sand-packed columns was firstly investigated under various conditions. The stable plateau values (C(max)) of the breakthrough curves (BTCs), critical PVs (the number of pore volumes
Guozhong Wu et al.
Chemosphere, 92(11), 1465-1471 (2013-05-02)
The molecular scale sorption, diffusion and distribution of asphaltene, resin, aromatic and saturate fractions of heavy crude oil on quartz surface were studied using molecular dynamic simulation. Sorption of saturates on quartz decreased by 31% when temperature increased from 298
Louise B Wright et al.
Physical chemistry chemical physics : PCCP, 15(13), 4715-4726 (2013-02-21)
Harnessing the properties of biomolecules, such as peptides, adsorbed on inorganic surfaces is of interest to many cross-disciplinary areas of science, ranging from biomineralisation to nanomedicine. Key to advancing research in this area is determination of the peptide conformation(s) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service