Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

28-1980

Sigma-Aldrich

Sodium bisulfite

SAJ first grade, ≥58.5% SO2 basis

Synonym(s):

Sodium hydrogensulfite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaHSO3
CAS Number:
Molecular Weight:
104.06
MDL number:
UNSPSC Code:
12352302
grade:
SAJ first grade
form:
solid

grade

SAJ first grade

form

solid

availability

available only in Japan

concentration

≥58.5% SO2

pH

4.3 (10 g/L)

storage temp.

15-25°C

SMILES string

[Na+].OS([O-])=O

InChI

1S/Na.H2O3S/c;1-4(2)3/h;(H2,1,2,3)/q+1;/p-2

InChI key

DWAQJAXMDSEUJJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

28-1980-5-500G-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah H Taylor et al.
BioTechniques, 52(4), 254-262 (2012-04-10)
RNA silencing has been exploited to produce transgenic plants with resistance to viral pathogens via posttranscriptional gene silencing (PTGS). In some cases, this technology is difficult to apply due to the instability of inverted repeat (IR) constructs during cloning and
J Y Zhu et al.
Bioresource technology, 102(19), 8921-8929 (2011-08-10)
Native aspen (Populus tremuloides) was pretreated using sulfuric acid and sodium bisulfite (SPORL) and dilute sulfuric acid alone (DA). Simultaneous enzymatic saccharification and fermentation (SSF) was conducted at 18% solids using commercial enzymes with cellulase loadings ranging from 6 to
Shuang-Qing Zhang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 880(1), 163-167 (2011-12-02)
A liquid chromatography-tandem mass spectrometry method for the determination of andrographolide sodium bisulphite (ASB) in rat urine was established and validated. To our knowledge, the analytical method is the first developed assay for the determination of ASB in urine samples.
Kequan Deng et al.
Molecules (Basel, Switzerland), 17(3), 2968-2979 (2012-03-13)
Defatted soybean flour was treated with hexane and ethanol to reduce lipid content and heated to inactivate lipoxygenase (LOX, linoleate:oxygen reductase; EC 1.13.11.12) to obtain lipid-reduced soybean flour (LRSF). The effects of processing conditions such as pH, reducing agent and
Miyako Yoshida et al.
Chemical & pharmaceutical bulletin, 61(1), 1-7 (2012-11-06)
The purpose of this study was to predict the stability of imipenem in a mixed infusion. The hydrolysis of imipenem in aqueous solution was found to be accelerated by pH, and by increasing concentrations of sodium bisulfite (SBS) and L-cysteine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service