Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

28-2430

Sigma-Aldrich

Sodium citrate tribasic dihydrate

SAJ first grade, ≥99.0%

Synonym(s):

Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COONa)(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
294.10
Beilstein:
6104939
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥99.0%
grade:
SAJ first grade
form:
solid

grade

SAJ first grade

Assay

≥99.0%

form

solid

availability

available only in Japan

pH

7.5-9.0 (25 °C, 29.4 g/L)

mp

>300 °C (lit.)

storage temp.

15-25°C

SMILES string

O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3

InChI key

NLJMYIDDQXHKNR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

28-2430-8-25KG-J:
28-2430-5-500G-J:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stijn Mertens et al.
Applied and environmental microbiology, 81(23), 8202-8214 (2015-09-27)
Lager beer is the most consumed alcoholic beverage in the world. Its production process is marked by a fermentation conducted at low (8 to 15°C) temperatures and by the use of Saccharomyces pastorianus, an interspecific hybrid between Saccharomyces cerevisiae and
Evgeny Smirnov et al.
ACS nano, 9(6), 6565-6575 (2015-06-04)
Soft or "liquid-liquid" interfaces were functionalized by roughly half a monolayer of mirror-like nanofilms of gold nanoparticles using a precise interfacial microinjection method. The surface coverage of the nanofilm was characterized by ion transfer voltammetry. These gold nanoparticle films represent
Ruben de Vries et al.
Journal of dairy science, 98(12), 8678-8687 (2015-10-12)
The aim of this study was to evaluate the effect of shortening or omitting the dry period of dairy cows on milk casein composition. For this study, we analyzed milk samples of 90 cows with a dry period of 0
Jun Zhao et al.
Nanotechnology, 25(40), 405101-405101 (2014-09-12)
The treatment of liver injuries or hepatocellular carcinoma (HCC) has been hindered by the lack of efficient drug delivery. Even with the help of nanoparticles or other synthetic delivering agents, a large portion of the dose is still sequestered in
Rasmus I Jølck et al.
Advanced healthcare materials, 3(10), 1680-1687 (2014-04-16)
External beam radiation therapy relies on the ability to deliver high radiation doses to tumor cells with minimal exposure to surrounding healthy tissue. Advanced irradiation techniques, including image-guided radiation therapy (IGRT), rely on the ability to locate tumors to optimize

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service