Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

C-41212

Sigma-Aldrich

DetachKit 2

3 x 125 ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

packaging

pkg of 3 × 125 ml

technique(s)

cell culture | mammalian: suitable

shipped in

cooler

storage temp.

−20°C

General description

DetachKit 2-125

Application

The DetachKit 2 was developed for gentle and effective detachment of adherent primary human cells in culture and is especially valuable for more sensitive cell types. Each batch is tested on primary human cells.

Components

Each DetachKit 2 consists of three components: HEPES BSS (HEPES buffered Balanced Salt Solution), Trypsin/EDTA Solution and TNS (Trypsin Neutralization Solution). The DetachKit 2 comes with a Lower Trypsin/EDTA ratio of 0.025%/0.01%. HEPES BSS contains 30 mM HEPES, D-Glucose, NaCl, KCl, Na-Phosphate and Phenol Red. TNS contains 0.05% Trypsin Inhibitor from soybean and 0.1% Bovine Serum Albumin.

Quality

All lots of PromoCell′s DetachKits are subjected to comprehensive quality control tests. Each lot is routinely tested for use with primary human cells, the absence of cytotoxicity, and physical parameters such as osmolality and pH level. Approved in-house lots are used as a reference. In addition, all lots have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Storage and Stability

Store at -20°C in the dark immediately after arrival. Refrigerate thawed solution at 4 to 8°C. If stored properly, the product is stable until the expiry date stated on the label. Thawed solutions can be stored at 4 to 8°C and used for up to 6 weeks.

Subculture Routine

Click here for more information.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Please refer to KIT Component information

PRTR

Please refer to KIT Component information

FSL

Please refer to KIT Component information

ISHL Indicated Name

Please refer to KIT Component information

ISHL Notified Names

Please refer to KIT Component information

Cartagena Act

Please refer to KIT Component information

JAN Code

キットコンポーネントの情報を参照してください


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service